missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hydroxypropafenone Hydrochloride, Mikromol™
Description
5-Hydroxypropafenone Hydrochloride, Mikromol
Specifications
Specifications
| Chemical Name or Material | 5-Hydroxypropafenone Hydrochloride |
| CAS | 86383-32-6 |
| Analyte or Component Names | 5-Hydroxypropafenone Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H27 N O4 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | 5-Hydroxypropafenone Hydrochloride |
| InChI Formula | InChI=1S/C21H27NO4.ClH/c1-2-12-22-14-18(24)15-26-21-11-9-17(23)13-19(21)20(25)10-8-16-6-4-3-5-7-16;/h3-7,9,11,13,18,22-24H,2,8,10,12,14-15H2,1H3;1H |
| Solution Type | Neat |
| SMILES | Cl.CCCNCC(O)COc1ccc(O)cc1C(=O)CCc2ccccc2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?