missing translation for 'onlineSavingsMsg'
Learn More

5-Iodosalicylic acid, 97%

Product Code. p-7025672
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11479197

Brand: Thermo Scientific Alfa Aesar B21291.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 119-30-2 | C7H5IO3 | 264.018 g/mol

2-hydroxy-5-iodobenzoic acid is a reagent in various organic chemical reactions leading to pharmaceutical compounds. It is involved in the synthesis of labeled benzamide analogs for tumor imaging as well as the synthesis of the dye, Congo Red, for biological analyses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-hydroxy-5-iodobenzoic acid is a reagent in various organic chemical reactions leading to pharmaceutical compounds. It is involved in the synthesis of labeled benzamide analogs for tumor imaging as well as the synthesis of the dye, Congo Red, for biological analyses.

Solubility
Solubility in methanol is almost transparent.

Notes
Light sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-30-2
Molecular Formula C7H5IO3
Molecular Weight (g/mol) 264.018
MDL Number MFCD00002458
InChI Key SWDNKOFGNPGRPI-UHFFFAOYSA-N
Synonym 5-iodosalicylic acid, benzoic acid, 2-hydroxy-5-iodo, salicylic acid, 5-iodo, 5-iodo-2-hydroxybenzoic acid, 2-hydroxy-5-iodo-benzoic acid, rarechem al be 0824, labotest-bb lt00454718, 5-jod-salicylsaure, 5-iodosalicylicacid, 5-iodo-salicylicaci
PubChem CID 8388
IUPAC Name 2-hydroxy-5-iodobenzoic acid
SMILES C1=CC(=C(C=C1I)C(=O)O)O

Specifications

Melting Point 194°C to 199°C
Quantity 25 g
Beilstein 2090443
Formula Weight 264.02
Percent Purity 97%
Chemical Name or Material 5-Iodosalicylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.