missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-1H-benzimidazole-2-thiol, Mikromol™

Product Code. 17973096
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17973096 25 mg 25mg
17931246 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17973096 Supplier Mikromol Supplier No. MM0095.030025
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-Methoxy-1H-benzimidazole-2-thiol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Methoxy-1H-benzimidazole-2-thiol
CAS 37052-78-1
Analyte or Component Names 5-Methoxy-1H-benzimidazole-2-thiol
Shipping Condition Room Temperature
Molecular Formula C8 H8 N2 O S
Certifications/Compliance ISO 17025
Synonym Esomeprazole Magnesium Trihydrate Imp. A (EP), Esomeprazole Imp. A (EP), Omeprazole Imp. A (EP), Omeprazole USP Related Compound B, Omeprazole USP RC B, 5-Methoxy-1H-benzimidazole-2-thiol, Omeprazole Related Compound B, Esomeprazole Magnesium Trihydrate Impurity A, Esomeprazole Sodium Impurity A, Omeprazole Impurity A, Esomeprazole Impurity A, Esomeprazole Impurity A
InChI Formula InChI=1S/C8H8N2OS/c1-11-5-2-3-6-7(4-5)10-8(12)9-6/h2-4H,1H3,(H2,9,10,12)
SMILES COc1ccc2[nH]c(S)nc2c1
IUPAC Name 5-methoxy-1H-benzimidazole-2-thiol
Molecular Weight (g/mol) 180.23
Quantity 25 mg
Formula Weight 180.0357 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.