missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-[[(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]sulphonyl]-1H-benzimidazole (Omeprazole Sulphone), Mikromol™

Product Code. 17922116
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17922116 25 mg 25mg
17920396 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17922116 Supplier Mikromol Supplier No. MM0095.050025
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-Methoxy-2-[[(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]sulphonyl]-1H-benzimidazole (Omeprazole Sulphone)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Omeprazole sulfone
CAS 88546-55-8
Analyte or Component Names Omeprazole sulfone
Shipping Condition Room Temperature
Molecular Formula C17 H19 N3 O4 S
Certifications/Compliance ISO 17025
Synonym Esomeprazole Magnesium Trihydrate Imp. D (EP), Esomeprazole Imp. D (EP), Omeprazole Imp. D (EP), Omeprazole USP Related Compound A, Omeprazole USP RC A, 5-Methoxy-2-[[(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]sulphonyl]-1H-benzimidazole, Omeprazole Sulphone, Omeprazole Related Compound A, Esomeprazole Magnesium Trihydrate Impurity D, Esomeprazole Sodium Impurity D, Omeprazole Impurity D, Esomeprazole Impurity D, Esomeprazole Impurity D
InChI Formula InChI=1S/C17H19N3O4S/c1-10-8-18-15(11(2)16(10)24-4)9-25(21,22)17-19-13-6-5-12(23-3)7-14(13)20-17/h5-8H,9H2,1-4H3,(H,19,20)
SMILES COc1ccc2[nH]c(nc2c1)S(=O)(=O)Cc3ncc(C)c(OC)c3C
IUPAC Name 5-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfonyl]-1H-benzimidazole
Molecular Weight (g/mol) 361.42
Quantity 25 mg
Formula Weight 361.1096 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.