missing translation for 'onlineSavingsMsg'
Learn More

5-Methylsalicylic acid, 98%

Product Code. 11410511
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410511 5 g 5g
11420511 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410511 Supplier Thermo Scientific Alfa Aesar Supplier No. B20040.06
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 89-56-5 | C8H8O3 | 152.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-56-5
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002461
InChI Key DLGBEGBHXSAQOC-UHFFFAOYSA-N
Synonym 5-methylsalicylic acid, p-cresotic acid, p-cresotinic acid, 6-hydroxy-m-toluic acid, p-homosalicylic acid, 2,5-cresotic acid, benzoic acid, 2-hydroxy-5-methyl, 5-methyl-2-hydroxybenzoic acid, 6-hydroxy-3-methylbenzoic acid, alpha-cresotinic acid
PubChem CID 6973
IUPAC Name 2-hydroxy-5-methylbenzoic acid
SMILES CC1=CC(=C(C=C1)O)C(=O)O

Specifications

Melting Point 151°C to 154°C
Odor Odorless
Quantity 5 g
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 5-Methylsalicylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.