missing translation for 'onlineSavingsMsg'
Learn More

5-Methylthiazol-2-amine, Mikromol™

Product Code. 17901906
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17901906 25 mg 25mg
17991906 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17901906 Supplier Mikromol Supplier No. MM0246.010025
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-Methylthiazol-2-amine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Methylthiazol-2-amine
CAS 7305-71-7
Analyte or Component Names 5-Methylthiazol-2-amine
Shipping Condition Room Temperature
Molecular Formula C4 H6 N2 S
Certifications/Compliance ISO 17025
Synonym Meloxicam Imp. B (EP), Meloxicam USP Related Compound B, Meloxicam USP RC B, 5-Methylthiazol-2-amine, Meloxicam Related Compound B, Meloxicam Impurity B
InChI Formula InChI=1S/C4H6N2S/c1-3-2-6-4(5)7-3/h2H,1H3,(H2,5,6)
SMILES Cc1cnc(N)s1
IUPAC Name 5-methyl-1,3-thiazol-2-amine
Molecular Weight (g/mol) 114.17
Quantity 25 mg
Formula Weight 114.0252 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.