missing translation for 'onlineSavingsMsg'
Learn More

5-Nitroso-6-ribitylamino-2,4(1H,3H)-pyrimidinedione Hydrochloride, TRC

Product Code. 30478131
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478131 25 mg 25mg
30465633 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30478131 Supplier Toronto Research Chemicals Supplier No. TRCN54661525MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Nitroso-6-ribitylamino-2,4(1H,3H)-pyrimidinedione Hydrochloride
Analyte or Component Names 5-Nitroso-6-ribitylamino-2,4(1H,3H)-pyrimidinedione Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C9H14N4O7 . HCl
Synonym 1-Deoxy-1-[(1,2,3,6-tetrahydro-5-nitroso-2,6-dioxo-4-pyrimidinyl)amino]-D-ribitol Hydrochloride
InChI Formula InChI=1S/C9H14N4O7.ClH/c14-2-4(16)6(17)3(15)1-10-7-5(13-20)8(18)12-9(19)11-7;/h3-4,6,14-17H,1-2H2,(H3,10,11,12,18,19);1H/t3-,4+,6-;/m0./s1
Solution Type Neat
SMILES O=C(C(N=O)=C(NC[C@@H]([C@H](O)[C@@H](CO)O)O)N1)NC1=O.Cl
IUPAC Name 5-nitroso-6-(((2S,3S,4R)-2,3,4,5-tetrahydroxypentyl)amino)pyrimidine-2,4(1H,3H)-dione hydrochloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 326.69
Quantity 25 mg
Formula Weight 326.07
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.