missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SLC22A8 Polyclonal specifically detects SLC22A8 in Human samples. It is validated for Immunohistochemistry/Immunohistochemistry (Paraffin).
Specifications
Specifications
| Antigen | SLC22A8 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | FITC |
| Formulation | PBS |
| Gene Alias | hOAT3, MGC24086, OAT3Organic anion transporter 3, solute carrier family 22 (organic anion transporter), member 8, solute carrier family 22 member 8 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human SLC22A8 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?