missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buspirone.HCl, Lipomed
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Buspirone.HCl
Specifications
Specifications
| Chemical Name or Material | Buspirone hydrochloride |
| CAS | 33386-08-2 |
| Analyte or Component Names | Buspirone hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H31 N5 O2 . Cl H |
| Synonym | Buspirone Hydrochloride |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.O=C1CC2(CCCC2)CC(=O)N1CCCCN3CCN(CC3)c4ncccn4 |
| IUPAC Name | 8-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]-8-azaspiro[4.5 ]decane-7,9-dione;hydrochloride |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction