missing translation for 'onlineSavingsMsg'
Learn More
Learn More
d,l-4-Methyl-N-ethyl-norephedrine.HCl, Lipomed
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
d,l-4-Methyl-N-ethyl-norephedrine.HCl
Specifications
Specifications
| Chemical Name or Material | d,l-4-Methyl-N-ethyl-norephedrine Hydrochloride |
| CAS | 1782625-25-5 |
| Analyte or Component Names | d,l-4-Methyl-N-ethyl-norephedrine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H19 N O . Cl H |
| Synonym | Benzenemethanol, α-[1-(ethylamino)ethyl]-4-methyl-, hydrochloride (1:1), α-[1-(Ethylamino)ethyl]-4-methylbenzenemethanol hydrochloride, (±)-4-Methyl-N-ethyl-norephedrine hydrochloride, 2-(Ethylamino)-1-(p-tolyl)propan-1-ol hydrochloride, 1-(4-Methylphenyl)-2-(ethylamino)propan-1-ol hydrochloride, (±)-1-(4-Methylphenyl)-2-ethylamino-1-propanol hydrochloride, (±)-1-(4-Methylphenyl)-2-ethylamino-1-propanol hydrochloride, (±)-1-(4-Methylphenyl)-2-ethylamino-1-propanol hydrochloride, (±)-1-(4-Methylphenyl)-2-ethylamino-1-propanol hydrochloride |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.CCNC(C)C(O)c1ccc(C)cc1 |
| IUPAC Name | 2-(ethylamino)-1-(4-methylphenyl)propan-1-ol;hydrochloride |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction