missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doxylamine.succinate, Lipomed
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Doxylamine.succinate
Specifications
Specifications
| Chemical Name or Material | Doxilamine succinate |
| CAS | 562-10-7 |
| Analyte or Component Names | Doxilamine succinate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H22 N2 O . C4 H6 O4 |
| Synonym | Doxylamine Hydrogen Succinate, Butanedioic acid, compd. with N,N-dimethyl-2-[1-phenyl-1-(2-pyridinyl)ethoxy]ethanamine (1:1), Ethanamine, N,N-dimethyl-2-[1-phenyl-1-(2-pyridinyl)ethoxy]-, butanedioate (1:1) (9 CI), Alsodorm, Decamium, Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI), Pyridine, 2-[alpha-[2-(dimethylamino)ethoxy]-alpha-methylbenzyl]-, succinate (1:1) (8 CI) |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | CN(C)CCOC(C)(c1ccccc1)c2ccccn2.OC(=O)CCC(=O)O |
| IUPAC Name | butanedioic acid;N,N-dimethyl-2-(1-phenyl-1-pyridin-2-ylethoxy)ethanamine |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction