missing translation for 'onlineSavingsMsg'
Learn More
Learn More
53BP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 96 publications
Brand: Novus Biologicals NB100-904SS
This item is not returnable.
View return policy
Description
53BP1 Polyclonal specifically detects 53BP1 in Human, Mouse, Other samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Proximity Ligation Assay, In-situ Hybridization, Knockdown Validated.
Specifications
| 53BP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| 53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and mouse. Human has been tested in WB, IHC and ICC/IF, mouse has only been tested in ICC/IF. | |
| Store at -20 degreesC. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), In situ PLA, In Situ Hybridization (ISH), KnockDown | |
| 1 mg/mL | |
| Western Blot 1:1000 - 1:10000, Flow Cytometry 2 - 5 ug/mL, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence, Immunoprecipitation reported in scientific literature (PMID 25512560), Immunohistochemistry-Paraffin 1:400, Proximity Ligation Assay reported in scientific literature (PMID 26734725), In-situ Hybridization reported in scientific literature (PMID 25407517), KnockDown Validated reported in scientific literature (PMID 31635359) | |
| Q12888 | |
| TP53BP1 | |
| 53BP1 Antibody was made to a synthetic peptide corresponding to a portion of human 53BP1 encoded in exon 11, 12 and 19 [Uniprot: Q12888]. This antibody is a cocktail hybrid of NB100-304 and NB100-305. | |
| 0.025 mL | |
| Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Tumor Suppressors | |
| 7158 | |
| Human, Mouse, Other | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction