missing translation for 'onlineSavingsMsg'
Learn More
Learn More
53BP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 96 publications
266.00 € - 575.00 €
Specifications
| Antigen | 53BP1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000 - 1:10000, Flow Cytometry 2 - 5 ug/mL, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence, Immunoprecipitation reported in scientific literature (PMID 25512560), Immunohistochemistry-Paraffin 1:400, Proximity Ligation Assay reported in scientific literature (PMID 26734725), In-situ Hybridization reported in scientific literature (PMID 25407517), KnockDown Validated reported in scientific literature (PMID 31635359) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), In situ PLA, In Situ Hybridization (ISH), KnockDown |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18787573
|
Novus Biologicals
NB100-904SS |
0.025 mL |
266.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18707103
|
Novus Biologicals
NB100-904 |
575.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
53BP1 Polyclonal specifically detects 53BP1 in Human, Mouse, Other samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Proximity Ligation Assay, In-situ Hybridization, Knockdown Validated.Specifications
| 53BP1 | |
| Western Blot 1:1000 - 1:10000, Flow Cytometry 2 - 5 ug/mL, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence, Immunoprecipitation reported in scientific literature (PMID 25512560), Immunohistochemistry-Paraffin 1:400, Proximity Ligation Assay reported in scientific literature (PMID 26734725), In-situ Hybridization reported in scientific literature (PMID 25407517), KnockDown Validated reported in scientific literature (PMID 31635359) | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Tumor Suppressors | |
| PBS with 0.02% Sodium Azide | |
| 53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1 | |
| TP53BP1 | |
| IgG | |
| Affinity Purified | |
| Human and mouse. Human has been tested in WB, IHC and ICC/IF, mouse has only been tested in ICC/IF. |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), In situ PLA, In Situ Hybridization (ISH), KnockDown | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Other | |
| Q12888 | |
| 7158 | |
| 53BP1 Antibody was made to a synthetic peptide corresponding to a portion of human 53BP1 encoded in exon 11, 12 and 19 [Uniprot: Q12888]. This antibody is a cocktail hybrid of NB100-304 and NB100-305. | |
| Primary | |
| Store at -20 degreesC. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title