missing translation for 'onlineSavingsMsg'
Learn More

5H-Dibenzo[b,f]azepine (Iminostilbene), Mikromol™

Product Code. 17971446
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17971446 100 mg 100mg
17903046 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17971446 Supplier Mikromol Supplier No. MM0076.08
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5H-Dibenzo[b,f]azepine (Iminostilbene)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iminostilbene
CAS 256-96-2
Analyte or Component Names Iminostilbene
Shipping Condition Room Temperature
Molecular Formula C14 H11 N
Certifications/Compliance ISO 17025
Synonym Carbamazepine Imp. D (EP), Oxcarbazepine Imp. E (EP), Carbamazepine USP Related Compound B, Carbamazepine USP RC B, 5H-Dibenzo[b,f]azepine, Iminostilbene, 2,2'-Iminostilbene, 5-Azadibenzo[a,e]cycloheptatriene, Dibenz[b,f]azepine, NSC 123458, RP 9989, Carbamazepine Related Compound B, Carbamazepine Impurity D, Oxcarbazepine Impurity E
InChI Formula InChI=1S/C14H11N/c1-3-7-13-11(5-1)9-10-12-6-2-4-8-14(12)15-13/h1-10,15H
SMILES N1c2ccccc2C=Cc3ccccc13
IUPAC Name 11H-benzo[b][1]benzazepine
Molecular Weight (g/mol) 193.24
Quantity 100 mg
Formula Weight 193.0891 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.