missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-3,4-dihydroquinolin-2(1H)-one, Mikromol™

Product Code. 17903516
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17903516 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17903516 Supplier Mikromol Supplier No. MM0961.130025
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

7-Hydroxy-3,4-dihydroquinolin-2(1H)-one
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,4-Dihydro-7-hydroxyquinoline-2(1H)-one
CAS 22246-18-0
Analyte or Component Names 3,4-Dihydro-7-hydroxyquinoline-2(1H)-one
Shipping Condition Room Temperature
Molecular Formula C9 H9 N O2
Certifications/Compliance ISO 17025
Synonym 7-Hydroxy-3,4-dihydroquinolin-2(1H)-one, 2(1H)-Quinolinone, 3,4-dihydro-7-hydroxy-, Carbostyril, 3,4-dihydro-7-hydroxy- (7CI,8CI), Hydrocarbostyril, 7-hydroxy- (6CI), 3,4-Dihydro-7-hydroxy-2(1H)-quinolinone, 7-Hydroxy-1,2,3,4-tetrahydro-2-quinolinone, 7-Hydroxy-3,4-dihydro-1H-quinolin-2-one, 7-Hydroxy-3,4-dihydro-2(1H)-quinolinone, 7-Hydroxy-3,4-dihydrocarbostyril, Aripiprazole Impurity A
InChI Formula InChI=1S/C9H9NO2/c11-7-3-1-6-2-4-9(12)10-8(6)5-7/h1,3,5,11H,2,4H2,(H,10,12)
SMILES Oc1ccc2CCC(=O)Nc2c1
IUPAC Name 7-hydroxy-3,4-dihydro-1H-quinolin-2-one
Molecular Weight (g/mol) 163.17
Quantity 25 mg
Formula Weight 163.0633 g/mol
Show More Show Less
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.