missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxyflavanone, 99%

Product Code. 11476396
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476396 1 g 1g
11496396 0.25 g 0.25g
11486396 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11476396 Supplier Thermo Scientific Alfa Aesar Supplier No. B24720.03
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 6515-36-2 | C15H12O3 | 240.258 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6515-36-2
Molecular Formula C15H12O3
Molecular Weight (g/mol) 240.258
MDL Number MFCD00017487
InChI Key SWAJPHCXKPCPQZ-UHFFFAOYSA-N
Synonym 7-hydroxyflavanone, 7-hydroxy-2-phenylchroman-4-one, chembl97542, 7-hydroxy-2-phenyl-4-chromanone, 7-hydroxy-2-phenyl-2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-7-hydroxy-2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2,3-dihydro-7-hydroxy-2-phenyl, 7-hydroxy-flavanone, d05zpo
PubChem CID 1890
ChEBI CHEBI:34483
IUPAC Name 7-hydroxy-2-phenyl-2,3-dihydrochromen-4-one
SMILES C1C(OC2=C(C1=O)C=CC(=C2)O)C3=CC=CC=C3

Specifications

Melting Point 187°C to 189°C
Quantity 1 g
Beilstein 87355
Formula Weight 240.25
Percent Purity 99%
Chemical Name or Material 7-Hydroxyflavanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.