missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxy-1-naphthalenol, TRC

Product Code. 30471266
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471266 1 g 1g
30481858 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30471266 Supplier Toronto Research Chemicals Supplier No. TRCM2647101G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67247-13-6
Molecular Formula C11 H10 O2
Molecular Weight (g/mol) 174.2
IUPAC Name 7-methoxynaphthalen-1-ol
SMILES COc1ccc2cccc(O)c2c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C11H10O2/c1-13-9-6-5-8-3-2-4-11(12)10(8)7-9/h2-7,12H,1H3
Formula Weight 174.0681
Chemical Name or Material 7-Methoxy-1-naphthalenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.