missing translation for 'onlineSavingsMsg'
Learn More

7-Nitro-1-tetralone, 97%

Product Code. 11301427
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11301427 2 g 2g
11311427 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11301427 Supplier Thermo Scientific Alfa Aesar Supplier No. L01853.04
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40353-34-2 | C10H9NO3 | 191.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40353-34-2
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD00019661
InChI Key GWAQYWSNCVEJMW-UHFFFAOYSA-N
Synonym 7-nitro-1-tetralone, 7-nitrotetralone, 7-nitro-3,4-dihydronaphthalen-1 2h-one, 1 2h-naphthalenone, 3,4-dihydro-7-nitro, 3,4-dihydro-7-nitro-1 2h-naphthalenone, 3,4-dihydro-7-nitronaphthalen-1 2h-one, wln: l66 bvt&j inw, 7-nitro-2,3,4-trihydronaphthalen-1-one, 1 2h-naphthalenone,4-dihydro-7-nitro, 7-nitro-tetralone
PubChem CID 38445
IUPAC Name 7-nitro-3,4-dihydro-2H-naphthalen-1-one
SMILES C1CC2=C(C=C(C=C2)[N+](=O)[O-])C(=O)C1

Specifications

Melting Point 103°C to 108°C
Quantity 2 g
Beilstein 1570515
Formula Weight 191.19
Percent Purity 97%
Chemical Name or Material 7-Nitro-1-tetralone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.