missing translation for 'onlineSavingsMsg'
Learn More

7H-Purin-6-amine Sulphate (Adenine Sulphate), Mikromol

Product Code. 30462519
Change view
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30462519 4 x 25 mg -
30051264 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30462519 Supplier Mikromol Supplier No. MM0784.0100254AMP
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

7H-Purin-6-amine Sulphate (Adenine Sulphate), Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Adenine Sulphate
CAS 321-30-2
Analyte or Component Names Adenine Sulphate
Shipping Condition Room Temperature
Molecular Formula 2[C5 H5 N5] . S O4 H2
Certifications/Compliance ISO 17025
Synonym Adenosine Imp. A (EP), 7H-Purin-6-amine Sulphate, Adenosine Imp. A (EP) as Sulphate, Adenine Sulphate, Adenosine Impurity A as Sulphate
InChI Formula InChI=1S/2C5H5N5.H2O4S/c2*6-4-3-5(9-1-7-3)10-2-8-4;1-5(2,3)4/h2*1-2H,(H3,6,7,8,9,10);(H2,1,2,3,4)
Solution Type Neat
SMILES Nc1ncnc2[nH]cnc12.Nc3ncnc4[nH]cnc34.OS(=O)(=O)O
IUPAC Name 9H-purin-6-amine;sulfuric acid
Molecular Weight (g/mol) 368.33
Quantity 4 x 25 mg
Formula Weight 368.0764 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.