missing translation for 'onlineSavingsMsg'
Learn More

8-Anilino-1-naphthalenesulfonic acid, 98%

Product Code. 11925251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11925251 25 g 25g
11915251 5 g 5g
11935251 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11925251 Supplier Thermo Scientific Acros Supplier No. 401210250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 82-76-8 | C16H13NO3S | 299.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82-76-8
Molecular Formula C16H13NO3S
Molecular Weight (g/mol) 299.34
InChI Key FWEOQOXTVHGIFQ-UHFFFAOYSA-N
Synonym 8-anilino-1-naphthalenesulfonic acid, phenylperi acid, n-phenyl peri acid, 8-phenylamino naphthalene-1-sulfonic acid, 1-anilino-8-naphthalenesulfonate, 1-anilino-8-naphthalenesulfonic acid, peri acid, phenyl, 1-naphthalenesulfonic acid, 8-phenylamino, 1-anilinonaphthalene-8-sulfonic acid, 8-anilino-1-naphthalene sulfonic acid
PubChem CID 1369
ChEBI CHEBI:39708
IUPAC Name 8-anilinonaphthalene-1-sulfonic acid
SMILES C1=CC=C(C=C1)NC2=CC=CC3=C2C(=CC=C3)S(=O)(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 183.0°C to 183.5°C
Color Black to Green
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 25 g
Merck Index 15,654
Formula Weight 299.34
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 8-Anilino-1-naphthalenesulfonic acid
Show More Show Less
compliance-icons
Product Identifier
  • 8-Anilino-1-naphthalenesulfonic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.