missing translation for 'onlineSavingsMsg'
Learn More

8-Fluoro-5-methyl-6-oxo-5,6-dihydro-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylic Acid, Mikromol™

Product Code. 17950716
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17950716 25 mg 25mg
17972896 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17950716 Supplier Mikromol Supplier No. MM0762.010025
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

8-Fluoro-5-methyl-6-oxo-5,6-dihydro-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8-Fluoro-5-methyl-6-oxo-5,6-dihydro-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylic Acid
CAS 84378-44-9
Analyte or Component Names 8-Fluoro-5-methyl-6-oxo-5,6-dihydro-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylic Acid
Shipping Condition Room Temperature
Molecular Formula C13 H10 F N3 O3
Certifications/Compliance ISO 17025
Synonym Flumazenil Imp. A (EP), Flumazenil USP Related Compound A, Flumazenil USP RC A, 8-Fluoro-5,6-dihydro-5-methyl-6-oxo-4H-imidazol-[1,5-alpha][1,4]benzodiazepine-3-carboxylic Acid, Flumazenil acid, Ro 15-3890, Flumazenil Related Compound A (USP), Flumazenil Related Compound A, Flumazenil Impurity A
InChI Formula InChI=1S/C13H10FN3O3/c1-16-5-10-11(13(19)20)15-6-17(10)9-3-2-7(14)4-8(9)12(16)18/h2-4,6H,5H2,1H3,(H,19,20)
SMILES CN1Cc2c(ncn2c3ccc(F)cc3C1=O)C(=O)O
IUPAC Name 8-fluoro-5-methyl-6-oxo-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylic acid
Molecular Weight (g/mol) 275.24
Quantity 25 mg
Formula Weight 275.0706 g/mol
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.