missing translation for 'onlineSavingsMsg'
Learn More

9,9'-(Hydrazine-1,2-diylidene)bis(9-(3,4-dimethyl-5-pentylfuran-2-yl)nonanoic Acid), TRC

Product Code. 30389554
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30389554 25 mg 25mg
30389260 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30389554 Supplier Toronto Research Chemicals Supplier No. TRCH68652025MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9,9'-(Hydrazine-1,2-diylidene)bis(9-(3,4-dimethyl-5-pentylfuran-2-yl)nonanoic Acid)
Analyte or Component Names 9,9'-(Hydrazine-1,2-diylidene)bis(9-(3,4-dimethyl-5-pentylfuran-2-yl)nonanoic Acid)
Shipping Condition Room Temperature
Molecular Formula C40 H64 N2 O6
InChI Formula InChI=1S/C40H64N2O6/c1-7-9-17-25-35-29(3)31(5)39(47-35)33(23-19-13-11-15-21-27-37(43)44)41-42-34(24-20-14-12-16-22-28-38(45)46)40-32(6)30(4)36(48-40)26-18-10-8-2/h7-28H2,1-6H3,(H,43,44)(H,45,46)/b41-33-,42-34-
Solution Type Neat
SMILES CCCCCc1oc(\C(=N/N=C(/CCCCCCCC(=O)O)\c2oc(CCCCC)c(C)c2C)\CCCCCCCC(=O)O)c(C)c1C
IUPAC Name (9Z)-9-[(Z)-[8-carboxy-1-(3,4-dimethyl-5-pentylfuran-2-yl)octylidene]hydrazinylidene]-9-(3,4-dimethyl-5-pentylfuran-2-yl)nonanoic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 668.946
Quantity 25 mg
Formula Weight 668.476
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.