missing translation for 'onlineSavingsMsg'
Learn More

9-Chloro-11beta,17-dihydroxy-16alpha-methyl-3,20-dioxopregna-1,4-dien-21-yl Acetate (Icometasone 21-Acetate), Mikromol™

Product Code. 18031710
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18031710 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18031710 Supplier Mikromol Supplier No. MM1272.140025
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

9-Chloro-11beta,17-dihydroxy-16alpha-methyl-3,20-dioxopregna-1,4-dien-21-yl Acetate (Icometasone 21-Acetate)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Icomethasone 21-Acetate
CAS 24916-91-4
Analyte or Component Names Icomethasone 21-Acetate
Shipping Condition Room Temperature
Molecular Formula C24 H31 Cl O6
Certifications/Compliance ISO 9001
Synonym Pregna-1,4-diene-3,20-dione, 21-(acetyloxy)-9-chloro-11,17-dihydroxy-16-methyl-, (11β,16α)- (9CI), Pregna-1,4-diene-3,20-dione, 9-chloro-11β,17,21-trihydroxy-16α-methyl-, 21-acetate (7CI,8CI), Icometasone-21-acetate, Icometasone 21-Acetate, Mometasone Furoate Imp. O (EP)
InChI Formula InChI=1S/C24H31ClO6/c1-13-9-18-17-6-5-15-10-16(27)7-8-21(15,3)23(17,25)19(28)11-22(18,4)24(13,30)20(29)12-31-14(2)26/h7-8,10,13,17-19,28,30H,5-6,9,11-12H2,1-4H3/t13-,17+,18+,19+,21+,22+,23+,24+/m1/s1
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(=O)C
IUPAC Name [2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-chloro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
Molecular Weight (g/mol) 450.95
Quantity 25 mg
Formula Weight 450.1809 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.