missing translation for 'onlineSavingsMsg'
Learn More

9-Chloro-11beta,21-dihydroxy-16beta-methyl-3,20-dioxopregna-1,4-dien-17-yl Propanoate (Beclometasone 17-Propionate), Mikromol™

Product Code. 30457637
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457637 4 x 25 mg -
18090990 25 mg 25mg
17909919 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457637 Supplier Mikromol Supplier No. MM0199.0100254AMP
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

9-Chloro-11beta,21-dihydroxy-16beta-methyl-3,20-dioxopregna-1,4-dien-17-yl Propanoate (Beclometasone 17-Propionate), Mikromol
TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Beclometasone 17-Propionate
CAS 5534-18-9
Analyte or Component Names Beclometasone 17-Propionate
Shipping Condition Room Temperature
Molecular Formula C25 H33 Cl O6
Certifications/Compliance ISO 17025
Synonym 9-Chloro-11beta,21-dihydroxy-16beta-methyl-3,20-dioxopregna-1,4-dien-17-yl Propanoate, Beclometasone Dipropionate Imp. H (EP), Beclometasone 17-Propionate, Beclomethasone 17-monopropionate, Beclomethasone 17-propionate, (11beta,16beta)-9-Chloro-11,21-dihydroxy-16-methyl-17-(1-oxopropoxy)pregna-1,4-diene-3,20-dione, 9-Chloro-11beta,17,21-trihydroxy-16beta-methylpregna-1,4-diene-3,20-dione 17-propionate, CCI 2382, Beclometasone Dipropionate Related Compound H, Beclometasone Dipropionate Impurity H
InChI Formula InChI=1S/C25H33ClO6/c1-5-21(31)32-25(20(30)13-27)14(2)10-18-17-7-6-15-11-16(28)8-9-22(15,3)24(17,26)19(29)12-23(18,25)4/h8-9,11,14,17-19,27,29H,5-7,10,12-13H2,1-4H3/t14-,17-,18-,19-,22-,23-,24-,25-/m0/s1
Solution Type Neat
SMILES CCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C)C(=O)CO
IUPAC Name [(8S,9R,10S,11S,13S,14S,16S,17R)-9-chloro-11-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] propanoate
Molecular Weight (g/mol) 464.98
Quantity 4 x 25 mg
Formula Weight 464.1966 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.