missing translation for 'onlineSavingsMsg'
Learn More

9-Chlorofluorene, TRC

Product Code. 30359019
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359019 1 g 1g
30364299 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30359019 Supplier Toronto Research Chemicals Supplier No. TRCC3662501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6630-65-5
Molecular Formula C13 H9 Cl
Molecular Weight (g/mol) 200.66
Synonym 9-Chloro-9H-fluorene (ACI), Fluorene, 9-chloro- (6CI, 7CI, 8CI), 9-Chlorofluorene, NSC 59976, 9H-Fluorene, 9-chloro- (9CI, ACI)
IUPAC Name 9-chloro-9H-fluorene
SMILES ClC1c2ccccc2c3ccccc13

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H9Cl/c14-13-11-7-3-1-5-9(11)10-6-2-4-8-12(10)13/h1-8,13H
Formula Weight 200.0393
Chemical Name or Material 9-Chlorofluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.