Learn More
A20/TNFAIP3 Antibody (59A426) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 41 publication
Brand: Novus Biologicals NBP1-77533
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
A20/TNFAIP3 Monoclonal specifically detects A20/TNFAIP3 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular), CyTOF-ready.
Tekniske data
| Immunohistochemistry (Paraffin), CyTOF | |
| Western Blot 2-4 ug/ml, Simple Western 1:100, Flow Cytometry 2.5 ug/ml, Immunohistochemistry 5 - 10 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunoprecipitation 1-2 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml. Use reported in scientific literature (Metellus (2010)), Flow (Intracellular) 2.5 ug/ml. Use reported in scientific literature (Hjelmeland (2010)), CyTOF-ready | |
| TNFAIP3 |
| Monoclonal | |
| A20TNFA1P2, EC 3.4.19.12, EC 6.3.2.-, MGC104522, MGC138687, OTU domain-containing protein 7C, OTUD7CMGC138688, Putative DNA-binding protein A20, TNF alpha-induced protein 3, tumor necrosis factor alpha-induced protein 3, tumor necrosis factor inducible protein A20, tumor necrosis factor, alpha-induced protein 3, Zinc finger protein A20 | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion