missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A20/TNFAIP3 Antibody (59A426) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 41 publication
237.00 € - 613.00 €
Specifications
| Antigen | A20/TNFAIP3 |
|---|---|
| Clone | 59A426 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2-4 ug/ml, Simple Western 1:100, Flow Cytometry 2.5 ug/ml, Immunohistochemistry 5 - 10 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunoprecipitation 1-2 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml. Use reported in scientific literature (Metellus (2010)), Flow (Intracellular) 2.5 ug/ml. Use reported in scientific literature (Hjelmeland (2010)), CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18797393
|
Novus Biologicals
NBP1-77533SS |
0.025 mg |
237.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18767153
|
Novus Biologicals
NBP1-77533 |
613.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
A20/TNFAIP3 Monoclonal specifically detects A20/TNFAIP3 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular), CyTOF-ready.Specifications
| A20/TNFAIP3 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| Mouse | |
| Cancer, Necroptosis, Signal Transduction | |
| P21580 | |
| 7128 | |
| Full length recombinant human A20/TNFAIP3 Antibody (59A426). The epitope has been mapped to the C-terminal portion of A20 (amino acids 440-790) | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 59A426 | |
| Western Blot 2-4 ug/ml, Simple Western 1:100, Flow Cytometry 2.5 ug/ml, Immunohistochemistry 5 - 10 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunoprecipitation 1-2 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml. Use reported in scientific literature (Metellus (2010)), Flow (Intracellular) 2.5 ug/ml. Use reported in scientific literature (Hjelmeland (2010)), CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| A20TNFA1P2, EC 3.4.19.12, EC 6.3.2.-, MGC104522, MGC138687, OTU domain-containing protein 7C, OTUD7CMGC138688, Putative DNA-binding protein A20, TNF alpha-induced protein 3, tumor necrosis factor alpha-induced protein 3, tumor necrosis factor inducible protein A20, tumor necrosis factor, alpha-induced protein 3, Zinc finger protein A20 | |
| TNFAIP3 | |
| IgG1 κ | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title