missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A20/TNFAIP3 Antibody (59A426) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 41 publication
Brand: Novus Biologicals NBP1-77533SS
This item is not returnable.
View return policy
Description
A20/TNFAIP3 Monoclonal specifically detects A20/TNFAIP3 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular), CyTOF-ready.
Specifications
| A20/TNFAIP3 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 2-4 ug/ml, Simple Western 1:100, Flow Cytometry 2.5 ug/ml, Immunohistochemistry 5 - 10 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunoprecipitation 1-2 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml. Use reported in scientific literature (Metellus (2010)), Flow (Intracellular) 2.5 ug/ml. Use reported in scientific literature (Hjelmeland (2010)), CyTOF-ready | |
| A20TNFA1P2, EC 3.4.19.12, EC 6.3.2.-, MGC104522, MGC138687, OTU domain-containing protein 7C, OTUD7CMGC138688, Putative DNA-binding protein A20, TNF alpha-induced protein 3, tumor necrosis factor alpha-induced protein 3, tumor necrosis factor inducible protein A20, tumor necrosis factor, alpha-induced protein 3, Zinc finger protein A20 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), CyTOF | |
| 59A426 | |
| Unconjugated | |
| P21580 | |
| TNFAIP3 | |
| Full length recombinant human A20/TNFAIP3 Antibody (59A426). The epitope has been mapped to the C-terminal portion of A20 (amino acids 440-790) | |
| 0.025 mg | |
| Cancer, Necroptosis, Signal Transduction | |
| 7128 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction