missing translation for 'onlineSavingsMsg'
Learn More

ABCA1 Antibody (5A1-1422.11) - BSA Free, Novus Biologicals™

Product Code. 18230971 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18230971 0.025 mL 25µL
18759613 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18230971 Supplier Novus Biologicals Supplier No. NB400165SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 1 publication

ABCA1 Monoclonal specifically detects ABCA1 in Mouse samples. It is validated for Flow Cytometry, Western Blot (Negative).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCA1
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 5A1-1422.11
Conjugate Unconjugated
Dilution Flow Cytometry 1:1000-1:3000, Western Blot (Negative)
Formulation Ascites with 0.1% Sodium Azide
Gene Alias ABC-1, ABC1ATP-binding cassette transporter A1, ATP-binding cassette 1, ATP-binding cassette transporter 1, ATP-binding cassette, sub-family A (ABC1), member 1, CERP, Cholesterol efflux regulatory protein, EC 2.7.7.8, EC 2.8.1.8, EC 3.6.3, EC 3.6.3.41, FLJ14958, HDLDT1, membrane-bound, MGC164864, MGC165011, TGDATP-binding cassette sub-family A member 1
Gene Symbols ABCA1
Host Species Rat
Immunogen Mouse ABCA1-expressing HeLa stable transfectants. [UniProt# P41233]
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Astrocyte Markers, Cancer, Cellular Markers, Cholesterol Metabolism, Glia Markers, Golgi Apparatus Markers, Lipid and Metabolism, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 19
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.