missing translation for 'onlineSavingsMsg'
Learn More

ABCB7 Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18673572 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18673572 0.1 mL 0.10mL
18642592 0.02 mL 0.02mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18673572 Supplier Novus Biologicals Supplier No. NBP2928640.1ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ABCB7 Polyclonal antibody specifically detects ABCB7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCB7
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias ABC transporter 7 protein, ABC7ATP-binding cassette sub-family B member 7, mitochondrial, ASATATP-binding cassette 7, Atm1p, ATP-binding cassette transporter 7, ATP-binding cassette, sub-family B (MDR/TAP), member 7, EC 3.6.3, EC 3.6.3.42, EST140535
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 503-753 of human ABCB7 (NP_004290.2). AIVGGSGSGKSTIVRLLFRFYEPQKGSIYLAGQNIQDVSLESLRRAVGVVPQDAVLFHNTIYYNLLYGNISASPEEVYAVAKLAGLHDAILRMPHGYDTQVGERGLKLSGGEKQRVAIARAILKDPPVILYDEATSSLDSITEETILGAMKDVVKHRTSIFIAHRLSTVVDADEIIVLDQGKVAERGTHHGLLANPHSIYSEMWHTQSSRVQNHDNPKWEAKKENISKEEERKKLQEEIVNSVKGCGNCSC
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 22
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.