missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABI3 Recombinant Rabbit Monoclonal Antibody (3V2G1)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558084
This item is not returnable.
View return policy
Description
Immunogen sequence: VPDGRLSAAS SAFSLASAGS AEGVGGAPTP KGQAAPPAPP LPSSLDPPPP PAAVEVFQRP PTLEELSPPP PDEELPLPLD LPPPPPLDGD ELGLPPPPPG FGPDEPSWVP ASYLEKVVTL YPYTSQKDNE LSFSEGTVIC VTRRYSDGWC EGVSSEGTGF FPGNYVEPSC.
Inhibits ectopic tumor cell metastasis of SRD cells. In vitro, reduces cell motility.
Specifications
| ABI3 | |
| Recombinant Monoclonal | |
| 1.08 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q9P2A4 | |
| ABI3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 197-366 of human ABI3 (NP_057512.2). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 3V2G1 | |
| Unconjugated | |
| ABI3 | |
| 2210414K06Rik; ABI family member 3; ABI family, member 3; ABI gene family member 3; ABI gene family, member 3; Abi3; AI987680; NESH; NESH protein; new molecule including SH3; SSH3BP3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 51225 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction