missing translation for 'onlineSavingsMsg'
Learn More

Abiraterone Acetate N-Oxide, Mikromol

Product Code. 30462932
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30462932 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462932 Supplier Mikromol Supplier No. MM3504.180025
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Abiraterone Acetate N-Oxide, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Abiraterone Acetate N-Oxide
CAS 2517964-85-9
Analyte or Component Names Abiraterone Acetate N-Oxide
Shipping Condition Room Temperature
Molecular Formula C26 H33 N O3
Certifications/Compliance ISO 9001
Synonym Abiraterone Acetate N-Oxide
InChI Formula InChI=1S/C26H33NO3/c1-17(28)30-20-10-12-25(2)19(15-20)6-7-21-23-9-8-22(18-5-4-14-27(29)16-18)26(23,3)13-11-24(21)25/h4-6,8,14,16,20-21,23-24H,7,9-13,15H2,1-3H3/t20-,21-,23-,24-,25-,26+/m0/s1
Solution Type Neat
SMILES CC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CC=C4c5ccc[n+]([O-])c5)[C@@H]3CC=C2C1
IUPAC Name [(3S,8R,9S,10R,13S,14S)-10,13-dimethyl-17-(1-oxidopyridin-1-ium-3-yl)-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
Molecular Weight (g/mol) 407.55
Quantity 25 mg
Formula Weight 407.246 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.