missing translation for 'onlineSavingsMsg'
Learn More

Abiraterone Isopropyl Ether, TRC

Product Code. 30136477
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136477 100 mg 100mg
30006439 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136477 Supplier Toronto Research Chemicals Supplier No. TRCA108530100MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2484719-15-3
Molecular Formula C27H37NO
Molecular Weight (g/mol) 391.59
Synonym 3-((3S,8R,9S,10R,13S,14S)-3-Isopropoxy-10,13-dimethyl-2,3,4,7,8,9,10,11,12,13,14,15-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl)pyridine
IUPAC Name 3-[(3S,8R,9S,10R,13S,14S)-10,13-dimethyl-3-propan-2-yloxy-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-17-yl]pyridine
SMILES [H][C@@]12CC=C3C[C@@H](OC(C)C)CC[C@]3(C)[C@@]1([H])CC[C@@]4(C)[C@@]2([H])CC=C4C5=CC=CN=C5

Specifications

Analyte or Component Names Abiraterone Isopropyl Ether
Shipping Condition Room Temperature
InChI Formula InChI=1S/C27H37NO/c1-18(2)29-21-11-13-26(3)20(16-21)7-8-22-24-10-9-23(19-6-5-15-28-17-19)27(24,4)14-12-25(22)26/h5-7,9,15,17-18,21-22,24-25H,8,10-14,16H2,1-4H3/t21-,22-,24-,25-,26-,27+/m0/s1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 100 mg
Chemical Name or Material Abiraterone Isopropyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.