Learn More
Invitrogen™ ACACB Recombinant Rabbit Monoclonal Antibody (24GB3230)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556950
 View more versions of this product
Product Code. 30275430
  • 146.00 € / 20µL
Description
ACACB Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Catalyzes the ATP-dependent carboxylation of acetyl-CoA to malonyl-CoA. Carries out three functions: biotin carboxyl carrier protein, biotin carboxylase and carboxyltransferase. Involved in inhibition of fatty acid and glucose oxidation and enhancement of fat storage. May play a role in regulation of mitochondrial fatty acid oxidation through malonyl-CoA-dependent inhibition of carnitine palmitoyltransferase 1.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ACACB
Recombinant Monoclonal
0.75 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
E9Q4Z2, O00763
ACACB
A synthesized peptide derived from human Acetyl-CoA Carboxylase (aa 350-450).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB3230
Unconjugated
ACACB
Acacb; Acc2; ACCB; ACC-beta; acetyl-CoA carboxylase 2; acetyl-CoA carboxylase beta; acetyl-Coenzyme A carboxylase 2; acetyl-Coenzyme A carboxylase beta; AI597064; AW743042; Biotin carboxylase; HACC275
Rabbit
Affinity chromatography
RUO
100705, 116719, 32
-20°C