missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACACB Recombinant Rabbit Monoclonal Antibody (24GB3230)

Rabbit Recombinant Monoclonal Antibody
146.00 € - 500.00 €
Specifications
| Antigen | ACACB |
|---|---|
| Clone | 24GB3230 |
| Concentration | 0.75 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275769
![]() |
Invitrogen™
MA556949 |
100 μL |
500.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275430
|
Invitrogen™
MA556950 |
20 μL |
146.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACACB Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Catalyzes the ATP-dependent carboxylation of acetyl-CoA to malonyl-CoA. Carries out three functions: biotin carboxyl carrier protein, biotin carboxylase and carboxyltransferase. Involved in inhibition of fatty acid and glucose oxidation and enhancement of fat storage. May play a role in regulation of mitochondrial fatty acid oxidation through malonyl-CoA-dependent inhibition of carnitine palmitoyltransferase 1.Specifications
| ACACB | |
| 0.75 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| E9Q4Z2, O00763 | |
| 100705, 116719, 32 | |
| A synthesized peptide derived from human Acetyl-CoA Carboxylase (aa 350-450). | |
| Antibody |
| 24GB3230 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| ACACB | |
| Acacb; Acc2; ACCB; ACC-beta; acetyl-CoA carboxylase 2; acetyl-CoA carboxylase beta; acetyl-Coenzyme A carboxylase 2; acetyl-Coenzyme A carboxylase beta; AI597064; AW743042; Biotin carboxylase; HACC275 | |
| ACACB | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title