missing translation for 'onlineSavingsMsg'
Learn More

Aceclofenac Isopropyl Ester, Mikromol™

Product Code. 18083390
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18083390 100 mg 100mg
17999679 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18083390 Supplier Mikromol Supplier No. MM0181.11
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Aceclofenac Isopropyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aceclofenac Isopropyl Ester
CAS 2514625-79-5
Analyte or Component Names Aceclofenac Isopropyl Ester
Shipping Condition Room Temperature
Molecular Formula C19 H19 Cl2 N O4
Certifications/Compliance ISO 9001
Synonym Aceclofenac Isopropyl Ester
InChI Formula InChI=1S/C19H19Cl2NO4/c1-12(2)26-18(24)11-25-17(23)10-13-6-3-4-9-16(13)22-19-14(20)7-5-8-15(19)21/h3-9,12,22H,10-11H2,1-2H3
SMILES CC(C)OC(=O)COC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl
IUPAC Name (2-oxo-2-propan-2-yloxyethyl) 2-[2-(2,6-dichloroanilino)phenyl]acetate
Molecular Weight (g/mol) 396.26
Quantity 100 mg
Formula Weight 395.0691 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.