missing translation for 'onlineSavingsMsg'
Learn More

Aceclofenac, Mikromol™

Product Code. 18072280
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18072280 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18072280 Supplier Mikromol Supplier No. MM0181.00
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Aceclofenac
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aceclofenac
CAS 89796-99-6
Analyte or Component Names Aceclofenac
Shipping Condition Room Temperature
Molecular Formula C16 H13 Cl2 N O4
Certifications/Compliance ISO 17034
Synonym Benzeneacetic acid, 2-[(2,6-dichlorophenyl)amino]-, carboxymethyl ester (9CI), 2-[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetoxy]acetic acid, Aceclofar, Airtal, Biofenac, Bristaflam, Falcol, Gerbin, PR 82/3, Tresquim, [[[[[[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetyl]oxy]acetyl]oxy]acetyl]oxy]acetic Acid (Diacetic Aceclofenac), [[[[[[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetyl]oxy]acetyl]oxy]acetyl]oxy]acetic Acid, Aceclofenac Imp. H (EP), Diacetic Aceclofenac, 2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid 2-[2-(carboxymethoxy)-2-oxoethoxy]-2-oxoethyl ester
InChI Formula InChI=1S/C16H13Cl2NO4/c17-11-5-3-6-12(18)16(11)19-13-7-2-1-4-10(13)8-15(22)23-9-14(20)21/h1-7,19H,8-9H2,(H,20,21)
SMILES OC(=O)COC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl
IUPAC Name 2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetic acid
Molecular Weight (g/mol) 354.18
Quantity 250 mg
Formula Weight 353.0222 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.