missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acemetacin, Mikromol™
Description
Acemetacin
Specifications
Specifications
| Chemical Name or Material | Acemetacin |
| CAS | 53164-05-9 |
| Analyte or Component Names | Acemetacin |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H18 Cl N O6 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Acemetacin, [[[1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]acetyl]oxy]acetic acid, 1H-Indole-3-acetic acid, 1-(4-chlorobenzoyl)-5-methoxy-2-methyl-, carboxymethyl ester1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indole-3-acetic acid carboxymethyl ester, 1-(p-Chlorobenzoyl)-5-methoxy-2-methylindole-3-acetic acid carboxymethyl ester, 2-[2-[1-[(4-Chlorophenyl)carbonyl]-5-methoxy-2-methylindol-3-yl]acetyloxy]acetic acid, Acemet, Acemetacin, Acemethacin, Acemethazine, Acemix, Emflex, K 708, Rantudil, Rheumibis, Rheutrop, Solart, TV 1322, TVX 1322 |
| InChI Formula | InChI=1S/C21H18ClNO6/c1-12-16(10-20(26)29-11-19(24)25)17-9-15(28-2)7-8-18(17)23(12)21(27)13-3-5-14(22)6-4-13/h3-9H,10-11H2,1-2H3,(H,24,25) |
| SMILES | COc1ccc2c(c1)c(CC(=O)OCC(=O)O)c(C)n2C(=O)c3ccc(Cl)cc3 |
| IUPAC Name | 2-[2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetyl]oxyacetic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?