missing translation for 'onlineSavingsMsg'
Learn More

Acesulfame Potassium, Mikromol

Product Code. 30461542
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30461542 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30461542 missing translation for 'mfr' Mikromol missing translation for 'supplierNo' MM1093.000250
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Acesulfame Potassium, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acesulfame potassium
CAS 55589-62-3
Analyte or Component Names Acesulfame potassium
Shipping Condition Room Temperature
Molecular Formula C4 H4 N O4 S . K
Certifications/Compliance ISO 17034
Synonym Acesulfame potassium, Potassium 6-methyl-1,2,3-oxathiazin-4-olate 2,2-dioxide, 1,2,3-Oxathiazin-4(3H)-one, 6-methyl-, 2,2-dioxide, potassium salt (1:1), 1,2,3-Oxathiazin-4(3H)-one, 6-methyl-, 2,2-dioxide, potassium salt (9CI), 6-Methyl-3,4-dihydro-1,2,3-oxathiazin-4-one 2,2-dioxide potassium salt, Acesulfame K, E 950, Otizon, Potassium 3,4-dihydro-6-methyl-1,2,3-oxathiazine-4-one-2,2-dioxide, Potassium acesulfamate, Potassium acesulfame, Sunett, Sunett D, Sunnett, Sweet One
InChI Formula InChI=1S/C4H5NO4S.K/c1-3-2-4(6)5-10(7,8)9-3;/h2H,1H3,(H,5,6);/q;+1/p-1
Solution Type Neat
SMILES [K+].CC1=CC(=O)[N-]S(=O)(=O)O1
IUPAC Name potassium;6-methyl-2,2-dioxo-1-oxa-2λ^{6}-thia-3-azanidacyclohex-5-en-4-one
Molecular Weight (g/mol) 201.24
Quantity 250 mg
Formula Weight 200.9498 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.