missing translation for 'onlineSavingsMsg'
Learn More

Acetanilide (Acetylaniline), TRC

Product Code. 30133801
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133801 10 mg 10mg
30008244 100 mg 100mg
17994775 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133801 Supplier Toronto Research Chemicals Supplier No. TRCA16833010MG
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-84-4
Molecular Formula C8 H9 N O
Molecular Weight (g/mol) 135.16
Synonym N-Phenylacetamide, Paracetamol Imp. D (EP), Acetanilide, Acetamidobenzene, Acetanil, Acetoanilide, Acetylaniline, Antifebrin, Benzenamine, N-acetyl-, N-Acetylaminobenzene, N-Acetylaniline, NSC 203231, NSC 7636, Phenalgene, Phenalgin
IUPAC Name N-phenylacetamide
SMILES CC(=O)Nc1ccccc1

Specifications

Analyte or Component Names N-Phenylacetamide
Shipping Condition Room Temperature
InChI Formula InChI=1S/C8H9NO/c1-7(10)9-8-5-3-2-4-6-8/h2-6H,1H3,(H,9,10)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 10 mg
Formula Weight 135.0684
Percent Purity >95%
Chemical Name or Material N-Phenylacetamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.