missing translation for 'onlineSavingsMsg'
Learn More

Acetanilide (Acetylaniline), TRC

Product Code. 30008244
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008244 100 mg 100mg
30133801 10 mg 10mg
17994775 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008244 Supplier Toronto Research Chemicals Supplier No. TRCA168330100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-84-4
Formula Weight 135.0684
InChI Formula InChI=1S/C8H9NO/c1-7(10)9-8-5-3-2-4-6-8/h2-6H,1H3,(H,9,10)
IUPAC Name N-phenylacetamide
Molecular Formula C8 H9 N O
Molecular Weight (g/mol) 135.16
Percent Purity >95%
SMILES CC(=O)Nc1ccccc1
Synonym N-Phenylacetamide, Paracetamol Imp. D (EP), Acetanilide, Acetamidobenzene, Acetanil, Acetoanilide, Acetylaniline, Antifebrin, Benzenamine, N-acetyl-, N-Acetylaminobenzene, N-Acetylaniline, NSC 203231, NSC 7636, Phenalgene, Phenalgin
Chemical Name or Material N-Phenylacetamide
Grade HPLC
Show More Show Less

Specifications

Analyte or Component Names N-Phenylacetamide
Shipping Condition Room Temperature
Solution Type Neat
Industry Type Pharmaceutical
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.