missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ACF1 Polyclonal antibody specifically detects ACF1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | ACF1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | CoraFluor 1 |
| Formulation | PBS |
| Gene Alias | ACF1hWALp1, ATP-dependent chromatin remodeling protein, ATP-dependent chromatin-remodeling protein, ATP-utilizing chromatin assembly and remodeling factor 1, bromodomain adjacent to zinc finger domain protein 1A, bromodomain adjacent to zinc finger domain, 1A, CHRAC subunit ACF1, hACF1FLJ14383, WALp1, WCRF180DKFZp586E0518, Williams syndrome transcription factor-related chromatin-remodeling factor 180 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1347-1556 of human ACF1 (NP_038476.2).,, Sequence:, VFVELLSPRRKRRGRKSANNTPENSPNFPNFRVIATKSSEQSRSVNIASKLSLQESESKRRCRKRQSPEPSPVTLGRRSSGRQGGVHELSAFEQLVVELVRHDDSWPFLKLVSKIQVPDYYDIIKKPIALNIIREKVNKCEYKLASEFIDDIELMFSNCFEYNPRNTSEAKAGTRLQAFFHIQAQKLGLHVTPSNVDQVSTPPAAKKSRI |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?