missing translation for 'onlineSavingsMsg'
Learn More

ACSL5 Antibody (CL0275) [DyLight 755], Novus Biologicals™

Product Code. 30297262 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30297262 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30297262 Supplier Novus Biologicals Supplier No. NBP252899IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ACSL5 Monoclonal antibody specifically detects ACSL5 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSL5
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL0275
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias ACS2, ACS5FACL5 for fatty acid coenzyme A ligase 5, acyl-CoA synthetase long-chain family member 5, EC 6.2.1, EC 6.2.1.3, FACL5fatty acid coenzyme A ligase 5, fatty-acid-Coenzyme A ligase, long-chain 5, LACS 5, Long-chain acyl-CoA synthetase 5, long-chain fatty acid coenzyme A ligase 5, long-chain-fatty-acid--CoA ligase 5
Host Species Mouse
Immunogen This antibody was developed using a recombinant protein derived from Q9ULC5, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 51703
Target Species Human
Content And Storage Store at 4⪚C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.