missing translation for 'onlineSavingsMsg'
Learn More

Adenine, LoGiCal

Product Code. 30461337
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461337 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461337 Supplier LoGiCal Supplier No. LGCFOR1495.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Adenine
CAS 73-24-5
Analyte or Component Names Adenine
Shipping Condition Room Temperature
Molecular Formula C5 H5 N5
Certifications/Compliance ISO 9001
Synonym Adenine, 7H-purin-6-amine
InChI Formula InChI=1S/C5H5N5/c6-4-3-5(9-1-7-3)10-2-8-4/h1-2H,(H3,6,7,8,9,10)
Solution Type Neat
SMILES Nc1ncnc2nc[nH]c12
IUPAC Name 7H-purin-6-amine
Industry Type Clinical Testing
Molecular Weight (g/mol) 135.13
Quantity 10 mg
Formula Weight 135.0545 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.