missing translation for 'onlineSavingsMsg'
Learn More

Adenosine 5'-Monophosphate Disodium Salt, LoGiCal

Product Code. 30462308
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30462308 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462308 Supplier LoGiCal Supplier No. LGCFOR3619.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Adenosine 5'-Monophosphate Disodium Salt
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Adenosine 5'-Monophosphate Disodium Salt
CAS 4578-31-8
Analyte or Component Names Adenosine 5'-Monophosphate Disodium Salt
Shipping Condition Room Temperature
Molecular Formula C10 H12 N5 O7 P . 2 Na
Certifications/Compliance ISO 9001
Synonym 5'-Adenylic acid sodium salt (1:2), 5'-Adenylic acid, disodium salt (7 CI,8 CI,9 CI), 5'-AMP disodium salt, AMP disodium salt, Adenosine 5'-monophosphate disodium, Disodium 9-(beta-D-ribofuranosyl)adenine 5'-monophosphate, Disodium 9-(beta-D-ribofuranosyl)adenine 5'-monophosphate, Disodium 9-(beta-D-ribofuranosyl)adenine 5'-monophosphate, Disodium 9-(beta-D-ribofuranosyl)adenine 5'-monophosphate, Disodium 9-(beta-D-ribofuranosyl)adenine 5'-monophosphate, Disodium 9-(beta-D-ribofuranosyl)adenine 5'-monophosphate, Disodium 9-(beta-D-ribofuranosyl)adenine 5'-monophosphate, Disodium 9-(beta-D-ribofuranosyl)adenine 5'-monophosphate
InChI Formula InChI=1 S/C10H14N5O7P0.2 Na/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(22-10)1-21-23(18,19)20;;/h2-4,6-7,10,16-17 H,1H2,(H2,11,12,13)(H2,18,19,20);;/q;2*+1/p-2/t4-,6-,7-,10-;;/m1../s1
SMILES [Na+].[Na+].Nc1ncnc2c1ncn2[C@@H]3 O[C@H](COP(=O)([O-])[O-])[C@@H](O)[C@H]3 O
IUPAC Name disodium;[(2 R,3 S,4 R,5 R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
Industry Type Forensics
Molecular Weight (g/mol) 391.18
Quantity 10 mg
Formula Weight 391.027 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.