missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine A2aR Antibody (7F6-G5-A2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-39474SS
This item is not returnable.
View return policy
Description
Adenosine A2aR Monoclonal antibody specifically detects Adenosine A2aR in Human, Mouse, Rat, Porcine, Canine, Guinea Pig, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow (Intracellular), CyTOF
Specifications
| Adenosine A2aR | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1-5 μg/mL, Immunohistochemistry 1 -5 μg/mL, Immunocytochemistry/ Immunofluorescence 1:50-1:100. Use reported in scientific literature (PMID 25836920), Immunohistochemistry-Paraffin 1 -5 μg/mL, Immunohistochemistry-Frozen reported in scientific literature (PMID 9822147), Flow (Intracellular) 1 - 5 μg/mL, CyTOF-ready | |
| P29274, P30543, Q60613 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Canine | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 7F6-G5-A2 | |
| Unconjugated | |
| PBS | |
| adenosine A2 receptor, adenosine A2a receptor, adenosine receptor A2a, adenosine receptor subtype A2a, ADORA 2, ADORA2, hA2aR, RDC8 | |
| Full length recombinant human Adenosine A2a R. [UniProt# P29274] | |
| 0.025 mL | |
| Apoptosis, GPCR, Immunology, Innate Immunity | |
| 135 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction