missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine A2aR Antibody (7F6-G5-A2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
275.00 € - 640.00 €
Specifications
| Antigen | Adenosine A2aR |
|---|---|
| Clone | 7F6-G5-A2 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1-5 μg/mL, Immunohistochemistry 1 -5 μg/mL, Immunocytochemistry/ Immunofluorescence 1:50-1:100. Use reported in scientific literature (PMID 25836920), Immunohistochemistry-Paraffin 1 -5 μg/mL, Immunohistochemistry-Frozen reported in scientific literature (PMID 9822147), Flow (Intracellular) 1 - 5 μg/mL, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18542621
|
Novus Biologicals
NBP1-39474SS |
0.025 mL |
275.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18503122
|
Novus Biologicals
NBP1-39474 |
0.1 mL |
640.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Adenosine A2aR Monoclonal antibody specifically detects Adenosine A2aR in Human, Mouse, Rat, Porcine, Canine, Guinea Pig, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow (Intracellular), CyTOFSpecifications
| Adenosine A2aR | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, GPCR, Immunology, Innate Immunity | |
| PBS | |
| adenosine A2 receptor, adenosine A2a receptor, adenosine receptor A2a, adenosine receptor subtype A2a, ADORA 2, ADORA2, hA2aR, RDC8 | |
| Full length recombinant human Adenosine A2a R. [UniProt# P29274] | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 7F6-G5-A2 | |
| Western Blot 1:1000, Flow Cytometry 1-5 μg/mL, Immunohistochemistry 1 -5 μg/mL, Immunocytochemistry/ Immunofluorescence 1:50-1:100. Use reported in scientific literature (PMID 25836920), Immunohistochemistry-Paraffin 1 -5 μg/mL, Immunohistochemistry-Frozen reported in scientific literature (PMID 9822147), Flow (Intracellular) 1 - 5 μg/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Pig, Canine | |
| P29274, P30543, Q60613 | |
| 135 | |
| IgG2a | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title