missing translation for 'onlineSavingsMsg'
Learn More

Adenovirus Antibody (8C4), Novus Biologicals™

Product Code. p-7106162 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
0.2 mg
Unit Size:
0.05mg
0.2mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18059053 0.05 mg 0.05mg
18716673 0.2 mg 0.2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18059053 Supplier Novus Biologicals Supplier No. NB600413SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 6 publications

Adenovirus Monoclonal specifically detects Adenovirus in Adenovirus, Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunodiffusion, Sandwich ELISA, Lateral Flow Assay, Block/Neutralize.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenovirus
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunodiffusion, ELISA, CyTOF, Neutralization
Classification Monoclonal
Clone 8C4
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunodiffusion, Sandwich ELISA 1:100-1:2000, CyTOF-ready, Block/Neutralize
Formulation PBS (pH 7.4) with 0.09% Sodium Azide
Gene Alias Adenovirus
Host Species Mouse
Immunogen Hexon antigen of human adenovirus.
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cell Biology, Stem Cell Markers, Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity Adenovirus hexon specific. The is directed against a genus-specific hexon antigen, which is present in all human serotypes. This detects a different epitope than the monoclonal, NB200-425.
Target Species Adenovirus, Virus
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.