missing translation for 'onlineSavingsMsg'
Learn More

ADP-ribosylarginine hydrolase Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18648814 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18648814 50 μL 50µL
18640374 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18648814 Supplier Novus Biologicals Supplier No. NBP32016850ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ADP-ribosylarginine hydrolase Polyclonal antibody specifically detects ADP-ribosylarginine hydrolase in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADP-ribosylarginine hydrolase
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias [Protein ADP-ribosylarginine] hydrolase, ADP-ribosylarginine hydrolaseARH1ADP-ribose-L-arginine cleaving enzyme, EC 3.2.2.19
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant human ADP-ribosylarginine hydrolase (Met1-Leu351; Accesion Number: NP_001116.1)
Purification Method Antigen and protein A Affinity-purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 141
Target Species Human
Content And Storage Store at 4&drg;C short term. Aliquot and store at -20&drg;C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.