missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adrenaline Tartrate (L-Adrenaline Tartrate), LoGiCal
Specifications
Specifications
| Chemical Name or Material | Adrenaline tartrate |
| CAS | 51-42-3 |
| Analyte or Component Names | Adrenaline tartrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H13 N O3 . C4 H6 O6 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Adrenaline Tartrate, L-Adrenaline Tartrate, 4-[(1R)-1-Hydroxy-2-(methylamino)ethyl]-1,2-benzenediol (2R,3R)-2,3-dihydroxybutanedioate (1:1) (salt), (R)-4-[1-Hydroxy-2-(methylamino)ethyl]-1,2-benzenediol [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1) (salt), Adrenaline tartrate (1:1), Tartaric acid compd. with adrenaline, (-)-Adrenaline acid tartrate, (-)-Adrenaline bitartrate, (-)-Adrenaline hydrogen tartrate, (-)-Adrenaline tartrate, (-)-Epinephrine (+)-bitartrate, (-)-Epinephrine bitartrate, Adrenalin bitartrate, Adrenaline acid tartrate, Adrenaline bitartrate, Adrenaline hydrogen tartrate, Asmatane Mist, Asthmahaler Epitrate, Bronitin Mist, Bronkaid Mist Suspension, Epinephrine bitartrate, Epinephrine d-bitartrate, Epinephrine hydrogen tartrate, Epinephrine tartrate, Epitrate, L-Adrenaline bitartrate, Medihaler-Epi, Primatene Mist Suspension, R-(-)-Epinephrine bitartrate, l-Adrenaline bitartrate, l-Adrenaline d-bitartrate, l-Adrenaline hydrogen tartrate, l-Adrenaline tartrate, l-Epinephrine bitartrate, l-Epinephrine d-bitartrate, (-)-3,4-Dihydroxy-alpha-[(methylamino)methyl]benzyl alcohol (+)-tartrate (1:1) (salt) |
| InChI Formula | InChI=1S/C9H13NO3.C4H6O6/c1-10-5-9(13)6-2-3-7(11)8(12)4-6;5-1(3(7)8)2(6)4(9)10/h2-4,9-13H,5H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t9-;1-,2-/m01/s1 |
| Solution Type | Neat |
| SMILES | CNC[C@H](O)c1ccc(O)c(O)c1.O[C@H]([C@@H](O)C(=O)O)C(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?